Rubidium ≥99.9%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Prescored ampoule.
93-3736-1G
93-3736-50G
93-3736-25G
93-3736-5G
100208-164EA
150.03
USD
100208-164
100203-458
100203-852
100204-716
Rubidium ≥99.9%
Rubidium
Formula:
Rb Boiling Pt: 688 °C Melting Pt: 38.9 °C Density: 1.532 |
MDL Number:
MFCD00134055 CAS Number: 7440-17-7 EINECS: 231-126-6 UN: 1423 ADR: 4.3,I Merck Index: 12,08439 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...