Rubidium carbonate ≥99.8% (Rb basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3725-5G
93-3725-25G
100199-082EA
60.86
USD
100199-082
100207-044
Rubidium carbonate ≥99.8% (Rb basis)
Rubidium carbonate
Formula:
CO₃Rb₂ MW: 230.94 g/mol Melting Pt: 837 °C Density: 2.43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011188 CAS Number: 584-09-8 EINECS: 209-530-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...