Rubidium hydroxide hydrate ≥99.8% (Rb basis), fused solid

Supplier: STREM CHEMICALS INC MS
Danger

93-3721-25G 93-3721-5G
100197-832EA 369.42 USD
100197-832 100197-834
Rubidium hydroxide hydrate ≥99.8% (Rb basis), fused solid
Rubidium hydroxide hydrate
Formula: H₃O₂Rb
MW: 120.49 g/mol
Melting Pt: 300…302 °C
Density: 3.203
MDL Number: MFCD00149835
CAS Number: 12026-05-0
EINECS: 215-186-0
UN: 2678
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR