Rubidium sulfate ≥99.8% (Rb basis)
Supplier: STREM CHEMICALS INC MS
93-3714-50G
93-3714-10G
100209-104EA
403.38
USD
100209-104
100197-828
Rubidium sulfate ≥99.8% (Rb basis)
Rubidium sulphate
Formula:
O₄Rb₂S Boiling Pt: ~1700 °C Melting Pt: 1050 °C Density: 3.613 Storage Temperature: Ambient |
MDL Number:
MFCD00011190 CAS Number: 7488-54-2 EINECS: 231-301-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...