Rubidium nitrate ≥99% (Rb basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-3711-50G 93-3711-10G
100196-634EA 290.15 USD
100196-634 100196-632
Rubidium nitrate ≥99% (Rb basis)
Rubidium nitrate
Formula: NO₃Rb
Boiling Pt: 578 °C (1013 hPa)
Melting Pt: 310 °C
Density: 3.11
Storage Temperature: Ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR