Rubidium iodide ≥99.8% (Rb basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-3709-5G 93-3709-25G
100199-080EA 52.37 USD
100199-080 100205-546
Rubidium iodide ≥99.8% (Rb basis)
Rubidium iodide
Formula: IRb
MW: 212.37 g/mol
Boiling Pt: 1300 °C
Melting Pt: 642 °C
Density: 3.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011192
CAS Number: 7790-29-6
EINECS: 232-198-1
Merck Index: 12,08443

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR