Rubidium hydroxide 50% in aqueous solution ≥99% (Rb basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-3708-100G 93-3708-25G
100209-102EA 1112.49 USD
100209-102 100205-544
Rubidium hydroxide 50% in aqueous solution ≥99% (Rb basis)
Rubidium hydroxide
Formula: RbOH
MW: 102.48 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 300…302 °C
Density: 1.74 g/cm³ (20 °C)
MDL Number: MFCD00011194
CAS Number: 1310-82-3
EINECS: 215-186-0
UN: 2677
ADR: 8,II
Merck Index: 12,08442

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR