Rubidium chloride ≥99% (Rb basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Rubidiumchloride

93-3705-10G 93-3705-50G
100208-554EA 104.74 USD
100208-554 100210-878
Rubidium chloride ≥99% (Rb basis)
Rubidium chloride
Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390 °C
Melting Pt: 715 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR