Rubidium chloride ≥99.8% (Rb basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Rubidiumchloride

93-3704-10G 93-3704-50G
100210-372EA 108.98 USD
100210-372 100205-878
Rubidium chloride ≥99.8% (Rb basis)
Rubidium chloride
Formula: RbCl
MW: 120.92 g/mol
Boiling Pt: 1390
Melting Pt: 715
Density: 2.80
Storage Temperature: Ambient
MDL Number: MFCD00011187
CAS Number: 7791-11-9
EINECS: 232-240-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR