Hydrobromic acid 48%

Supplier: STREM CHEMICALS INC MS
Danger

93-3504-5000G 93-3504-1000G
100202-794EA 816.68 USD
100202-794 100205-542
Hydrobromic acid 48%
Hydrobromic acid
Formula: HBr
MW: 80.91 g/mol
Boiling Pt: 126 °C
Melting Pt: –11 °C
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR