Selenium disulphide ≥94%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Selenium sulphide, Selenium(IV) sulphide

93-3415-25G 93-3415-100G
100202-102EA 72.18 USD
100202-102 100207-042
Selenium disulphide ≥94%
Selenium disulphide
Formula: S₂Se
MW: 143.09
Boiling Pt: -112 Dec.
Melting Pt: < 100 °C
Density: 0.03 g/cm³ (–65 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011227
CAS Number: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR