Selenium tetrabromide ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-3410-25G 93-3410-5G
100202-792EA 376.49 USD
100202-792 100205-534
Selenium tetrabromide ≥99%
Selenium tetrabromide
Formula: Br₄Se
Melting Pt: 75 °C (dec.)
Density: 4.029 g/cm³ (–78 °C)
MDL Number: MFCD00049766
CAS Number: 7789-65-3
EINECS: 232-181-9
UN: 3290
ADR: 6.1,II
Merck Index: 12,08581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR