Seleninyl dichloride ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

93-3408-25G 93-3408-5G
100197-820EA 451.51 USD
100197-820 100197-822
Seleninyl dichloride ≥97%
Seleninyl dichloride
Formula: Cl₂OSe
Boiling Pt: 176.4 °C
Melting Pt: 8.5 °C
Density: 2.44 g/cm³ (20 °C)
MDL Number: MFCD00016327
CAS Number: 7791-23-3
EINECS: 232-244-0
UN: 2879
ADR: 8,I
Merck Index: 12,08578

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR