Arsenic (III) sulfide ≥99.9% (As basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Arsenic trisulphide, Disrsenic trisulphide
93-3316-10G
93-3316-50G
100207-040EA
159.94
USD
100207-040
100200-128
Arsenic (III) sulfide ≥99.9% (As basis)
Arsenic(III) sulphide
Formula:
As₂S₃ MW: 246.04 g/mol Boiling Pt: 707 °C Melting Pt: 300 °C Density: 3.43 |
MDL Number:
MFCD00003435 CAS Number: 1303-33-9 EINECS: 215-117-4 UN: 1557 ADR: 6.1 II Merck Index: 12,00846 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...