Arsenic(III) oxide, Technical Grade
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
di-Arsenic trioxide
93-3313-50G
93-3313-250G
100205-530EA
48.12
USD
100205-530
100198-360
Arsenic(III) oxide, Technical Grade
Arsenic(III) oxide
Formula:
As₂O₃ MW: 197.84 g/mol Boiling Pt: 465 (subl.) Melting Pt: 312 Density: 3.738 Storage Temperature: Ambient |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1 II Merck Index: 12,00844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...