Arsenic(III) iodide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

93-3312-25G 93-3312-100G
100197-224EA 830.83 USD
100197-224 100208-550
Arsenic(III) iodide ≥98%
Arsenic(III) iodide
Formula: AsI₃
MW: 455.64 g/mol
Boiling Pt: 403 °C
Melting Pt: 146 °C
Density: 4.39 g/cm³ (20 °C)
MDL Number: MFCD00014170
CAS Number: 7784-45-4
EINECS: 232-068-4
UN: 1557
ADR: 6.1,II
Merck Index: 12,00843

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR