Arsenic(III) chloride ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-3310-100G 93-3310-25G
100207-834EA 768.55 USD
100207-834 100198-358
Arsenic(III) chloride ≥99%
Arsenic(III) chloride
Formula: AsCl₃
Boiling Pt: 130.2 °C
Melting Pt: –8.5 °C
Density: 2.163
MDL Number: MFCD00014165
CAS Number: 7784-34-1
EINECS: 232-059-5
UN: 1560
ADR: 6.1,I
Merck Index: 12,00841

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR