Diarsenic triselenide ≥99.999% (As basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Arsenic(III) selenide

93-3305-5G 93-3305-25G
100204-704EA 158.52 USD
100204-704 100206-148
Diarsenic triselenide ≥99.999% (As basis), PURATREM
Diarsenic triselenide
Formula: As₂Se₃
MW: 386.72 g/mol
Melting Pt: ~360 °C
Density: 4.49 g/cm³ (20 °C)
MDL Number: MFCD00014166
CAS Number: 1303-36-2
EINECS: 215-119-5
UN: 3077
ADR: 9,III
Merck Index: 12,00845

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR