Germanium ≥99.9999%, powder

Supplier: STREM CHEMICALS INC MS
Warning

93-3230-5G 93-3230-25G
100205-524EA 186.83 USD
100205-524 100200-126
Germanium ≥99.9999%, powder
Germanium
Formula: Ge
Boiling Pt: 2830 °C
Melting Pt: 937.4 °C
Density: 5.323
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR