Germanium(IV) iodide ≥99.999% (Ge basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tetraiodogermane, Germanium tetraiodide
93-3226-5G
93-3226-25G
100202-094EA
198.15
USD
100202-094
100207-216
Germanium(IV) iodide ≥99.999% (Ge basis), PURATREM
Germanium(IV) iodide
Formula:
GeI₄ MW: 580.23 g/mol Boiling Pt: 350 °C (1013 hPa) Melting Pt: 146 °C Density: 4.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016120 CAS Number: 13450-95-8 EINECS: 236-613-7 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...