Germanium diselenide ≥99.999% (Ge basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Germanium(IV) selenide

93-3224-5G 93-3224-1G
100200-124EA 852.06 USD
100200-124 100210-874
Germanium diselenide ≥99.999% (Ge basis), PURATREM
Germanium diselenide
Formula: GeSe₂
MW: 230.53
Melting Pt: 707
Density: 4.56
MDL Number: MFCD00016116
CAS Number: 12065-11-1
EINECS: 235-063-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR