Germanium(II) iodide ≥99.99% (Ge basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Germanium diiodide

93-3205-1G 93-3205-5G
100196-624EA 227.88 USD
100196-624 100196-626
Germanium(II) iodide ≥99.99% (Ge basis), PURATREM
Germanium(II) iodide
Formula: GeI₂
MW: 326.42 g/mol
Melting Pt: Dec.
Density: 5.4 g/cm³ (20 °C)
MDL Number: MFCD00135537
CAS Number: 13573-08-5
EINECS: 236-998-1
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR