Gallium(III) chloride, anhydrous ≥99.999%, granular, PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Gallium trichloride

93-3140-100G 93-3140-25G
100201-276EA 485.48 USD
100201-276 100196-622
Gallium(III) chloride, anhydrous ≥99.999%, granular, PURATREM
Gallium(III) chloride
Formula: Cl₃Ga
MW: 176.08 g/mol
Boiling Pt: 201.3 °C
Melting Pt: 77.9 °C
Density: 2.29 g/cm³ (80 °C)
MDL Number: MFCD00011018
CAS Number: 13450-90-3
EINECS: 236-610-0
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR