Gallium(III) acetylacetonate ≥99.99%, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Tris(2,4-pentanedionato-O,O')gallium (III), Gallium (III) 2,4-pentanedionate, Gallium (III) tris(2,4-pentanedionato-O,O'), Gallium(III) Acetylacetonate, Acetylacetone Gallium(III) salt, Tris(2,4-pentanedionato)gallium(III), Ga(acac)3
93-3130-25G
93-3130-5G
100197-222EA
342.52
USD
100197-222
100200-552
Gallium(III) acetylacetonate ≥99.99%, PURATREM
Gallium(III) acetylacetonate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₅H₂₁GaO₆ MW: 367.05 Boiling Pt: 140 °C (10 mmHg) (subl.) Melting Pt: 192…194 °CC >280 °C (dec.) Density: 1.42 |
MDL Number:
MFCD00013492 CAS Number: 14405-43-7 EINECS: 238-377-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...