Gallium triiodide, anhydrous ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-3110-5G 93-3110-25G
100208-548EA 116.06 USD
100208-548 100208-546
Gallium triiodide, anhydrous ≥99%
Gallium triiodide
Formula: GaI₃
MW: 450.44
Boiling Pt: 345
Melting Pt: 212
Density: 4.15 g/cm³ (25 °C)
MDL Number: MFCD00016107
CAS Number: 13450-91-4
EINECS: 236-611-6
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR