Gallium tribromide, anhydrous ≥99.999%, granular, PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Gallium (III) bromide

93-3109-1G 93-3109-5G
100197-218EA 389.23 USD
100197-218 100207-290
Gallium tribromide, anhydrous ≥99.999%, granular, PURATREM
Gallium tribromide
Formula: Br₃Ga
MW: 309.44 g/mol
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 121.5 °C
Density: 3.69 g/cm³ (20 °C)
MDL Number: MFCD00016100
CAS Number: 13450-88-9
EINECS: 236-609-5
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR