Gallium trinitrate hydrate ≥99.99%, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-3105-25G
93-3105-5G
100200-334EA
268.92
USD
100200-334
100203-450
Gallium trinitrate hydrate ≥99.99%, PURATREM
Gallium trinitrate hydrate
Formula:
GaN₃O₉·H₂O MW: 273.75 Melting Pt: 110 °C (dec.) |
MDL Number:
MFCD00149723 CAS Number: 69365-72-6 EINECS: 236-815-5 UN: 1477 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...