Gallium(II) chloride, anhydrous ≥99.999%, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-3104-1G
93-3104-5G
100205-518EA
111.82
USD
100205-518
100199-552
Gallium(II) chloride, anhydrous ≥99.999%, PURATREM
Gallium(II) chloride
Formula:
Ga₂Cl₄ Boiling Pt: 535 °C Melting Pt: 164 °C |
MDL Number:
MFCD00151526 CAS Number: 13498-12-9 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...