Zinc ≥99.9%, Powder

Supplier: STREM CHEMICALS INC MS
Danger

93-3060-2000G 93-3060-500G
100207-288EA 434.52 USD
100207-288 100202-084
Zinc ≥99.9%, Powder
Zinc
Formula: Zn
Boiling Pt: 907 °C
Melting Pt: 419.5 °C
Density: 7,14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR