Zinc(II) telluride ≥99.99% (Zn basis), PURATREM

Supplier: STREM CHEMICALS INC MS

93-3046-50G 93-3046-10G
100210-868EA 774.22 USD
100210-868 100210-354
Zinc(II) telluride ≥99.99% (Zn basis), PURATREM
Zinc(II) telluride
Formula: TeZn
MW: 192.99 g/mol
Melting Pt: 1240 °C
Density: ~ 6.3 g/cm³ (20 °C)
MDL Number: MFCD00043079
CAS Number: 1315-11-3
EINECS: 215-260-2
Merck Index: 12,10297

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR