Zinc perchlorate hexahydrate 99

Supplier: STREM CHEMICALS INC MS
Danger

93-3018-500G 93-3018-100G
100199-550EA 159.94 USD
100199-550 100205-508
Zinc perchlorate hexahydrate 99
Zinc perchlorate hexahydrate
Formula: Cl₂O₈Zn·6H₂O
MW: 372.38
Boiling Pt: 200 °C (dec.)
Melting Pt: 106
Density: 2.252
MDL Number: MFCD00150370
CAS Number: 10025-64-6
EINECS: 237-122-0
UN: 1481
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR