Zinc oxide ≥99.7%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Zinci oxidum, Zinc (II) oxide
93-3017-2000G
93-3017-500G
100201-098EA
168.43
USD
100201-098
100208-542
Zinc oxide ≥99.7%
Zinc oxide
Formula:
ZnO MW: 81.39 g/mol Melting Pt: 1975 °C Density: 5.61 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 13,10200 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...