Zinc iodide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Zinc (II) iodide

93-3014-250G 93-3014-50G
100202-070EA 264.68 USD
100202-070 100202-072
Zinc iodide ≥98%
Zinc iodide
Formula: I₂Zn
MW: 319.2 g/mol
Boiling Pt: 620 (dec.)
Melting Pt: 446
Density: 4.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 1759
ADR: 8 II
Merck Index: 12,10269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR