Zinc fluoride tetrahydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-3012-250G 93-3012-50G
100199-548EA 251.94 USD
100199-548 100205-506
Zinc fluoride tetrahydrate ≥98%
Zinc fluoride tetrahydrate
Formula: F₂Zn·4H₂O
MW: 175.45
Melting Pt: 872
Density: 2.255
MDL Number: MFCD00149892
CAS Number: 13986-18-0
EINECS: 232-001-9
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR