Methylmagnesium bromide 3M in ether

Supplier: STREM CHEMICALS INC MS
Danger

93-1265-250MMOL 93-1265-1MOL
100196-486EA 86.34 USD
100196-486 100206-930
Methylmagnesium bromide 3M in ether
Methylmagnesium bromide

Grignard

Formula: CH₃BrMg
MW: 119.24 g/mol
MDL Number: MFCD00000041
CAS Number: 75-16-1
EINECS: 200-844-1
UN: 3399
ADR: 4.3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR