Tetraethylammonium tetrahydroborate

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tetraethylammonium borohydride, Tetraethylammonium tetrahydridoborate

93-0573-10G 93-0573-2G
100208-080EA 492.55 USD
100208-080 101419-154
Tetraethylammonium tetrahydroborate
Tetraethylammonium tetrahydroborate
Formula: C₈H₂₄BN
MW: 145.1 g/mol
Melting Pt: 225 °C (dec.)
Density: 0.926
MDL Number: MFCD00031600
CAS Number: 17083-85-1
EINECS: 241-140-4
UN: 3134
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR