Gold(I) chloride ≥97% (99.99%-Au), PURATREM

Supplier: STREM CHEMICALS INC MS

79-1900-1G 79-1900-250MG 79-1900-5G
100204-656EA 308.55 USD
100204-656 100208-966 100200-000
Gold(I) chloride ≥97% (99.99%-Au), PURATREM
Gold(I) chloride
Formula: AuCl
MW: 232.42 g/mol
Melting Pt: 170 (dec.)
Density: 7.4
MDL Number: MFCD00046175
CAS Number: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8 III
Merck Index: 12,04528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR