Tetrakis(triphenylphosphine)platinum(0) ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Tetrakis(triphenylphosphine)platinum
78-2030-5G
78-2030-250MG
78-2030-1G
100201-032EA
481.23
USD
100201-032
100210-096
100198-902
Tetrakis(triphenylphosphine)platinum(0) ≥98%
Tetrakis(triphenylphosphine)platinum(0)
Formula:
C₇₂H₆₀P₄Pt MW: 1244.24 |
MDL Number:
MFCD00010014 CAS Number: 14221-02-4 EINECS: 238-087-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...