Potassium tetracyanoplatinate(II) hydrate

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Potassium platinum(II) cyanide hydrate

78-1980-5G 78-1980-1G 78-1980-250MG
100205-994EA 922.83 USD
100205-994 100209-386 100200-492
Potassium tetracyanoplatinate(II) hydrate
Potassium tetracyanoplatinate(II) hydrate
Formula: K₂Pt(CN)₄·xH₂O
MW: 377.34 g/mol (anhy)
MDL Number: MFCD00149920
CAS Number: 38245-27-1
EINECS: 209-236-0
UN: 1588
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR