Platinium(IV) oxide hydrate (~80-81% Pt)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
78-1890-1G
78-1890-5G
78-1890-250MG
100208-472EA
276
USD
100208-472
100208-474
101442-446
Platinium(IV) oxide hydrate (~80-81% Pt)
Platinium(IV) oxide hydrate
Formula:
O₂Pt·H₂O Melting Pt: 450 °C Density: 10.2 g/cm³ (20 °C) |
MDL Number:
MFCD00066964 CAS Number: 52785-06-5 EINECS: 215-223-0 UN: 1479 ADR: 5.1,II Merck Index: 12,07686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...