Platinum ≥99.999%, powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Platinium
78-1800-2G
78-1800-500MG
100210-750EA
778.46
USD
100210-750
100206-836
Platinum ≥99.999%, powder
Platinum
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Density: 21.45 Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 UN: 3089 ADR: 4.1 III Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...