Platinum ≥99.95%, foil

Supplier: STREM CHEMICALS INC MS

Synonyms: Platinium

78-0050-25X25MM 78-0050-50X50MM
100206-806EA 774.22 USD
100206-806 100209-272
Platinum ≥99.95%, foil
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR