Iridium ≥99.95%, sponge
Supplier: STREM CHEMICALS INC MS
|
Warning
|
77-2580-500MG
77-2580-2G
100197-604EA
123.14
USD
100197-604
100211-168
Iridium ≥99.95%, sponge
Iridium
Formula:
Ir MW: 192.22 g/mol Boiling Pt: 4130 °C Melting Pt: 2410 °C Density: 22,421 |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...