Iridium ≥99.8%, powder

Supplier: STREM CHEMICALS INC MS
Warning

77-2500-500MG 77-2500-2G
100197-602EA 283.08 USD
100197-602 100211-166
Iridium ≥99.8%, powder
Iridium
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 4500 °C (1013 hPa)
Melting Pt: 2443 °C
Density: 22.65 g/cm³ (20 °C)
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III
Merck Index: 12,05098

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR