Iridium ≥99.8%, foil

Supplier: STREM CHEMICALS INC MS

77-2090-25X25MM 77-2090-50X50MM
100210-086EA 935.57 USD
100210-086 100210-084
Iridium ≥99.8%, foil
Iridium
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 5300 °C (1013 hPa)
Melting Pt: 2450 °C
Density: 22,421
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR