Rhenium ≥99.99%, foil
Supplier: STREM CHEMICALS INC MS
|
Warning
|
75-0060-25X25MM
75-0060-50X50MM
100198-876EA
366.58
USD
100198-876
100206-796
Rhenium ≥99.99%, foil
Rhenium
Formula:
Re MW: 186.21 g/mol Boiling Pt: 5630 °C (1013 hPa) Melting Pt: 3180 °C Density: 21.02 g/cm³ (20 °C) |
MDL Number:
MFCD00011195 CAS Number: 7440-15-5 EINECS: 231-124-5 Merck Index: 12,08343 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...