Lutetium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS
Warning

71-0050-25X25MM 71-0050-50X50MM
100197-028EA 3586.58 USD
100197-028 100210-748
Lutetium ≥99.9%, foil
Lutetium
Formula: Lu
MW: 174.97 g/mol
Boiling Pt: 3327 °C
Melting Pt: 1652 °C
Density: 9.849 g/cm³ (25 °C)
MDL Number: MFCD00011098
CAS Number: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR