Cadmium acetate, anhydrous ≥99.999% (Cd basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

48-0100-5G 48-0100-25G
101419-098EA 162.77 USD
101419-098 101419-096
Cadmium acetate, anhydrous ≥99.999% (Cd basis), PURATREM
Cadmium acetate
Formula: (H₃CCOO)₂Cd
MW: 230.5 g/mol
Melting Pt: 256 °C
Density: 2.34 g/cm³ (20 °C)
MDL Number: MFCD00012449
CAS Number: 543-90-8
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III
Merck Index: 12,01650

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR