Palladium(II) acetate ≥99% (99.95+%-Pd)

Supplier: STREM CHEMICALS INC MS

Synonyms: Palladium diacetate, Palladium (II) acetate

46-1781-5G 46-1781-1G
101419-078EA 682.22 USD
101419-078 101419-076
Palladium(II) acetate ≥99% (99.95+%-Pd)
Palladium(II) acetate
Formula: C₄H₆O₄Pd
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR