Pentacarbonyliron ≥99.5% (99.9+%-Fe)

Supplier: STREM CHEMICALS INC MS
Danger


Packed in a Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC).

26-2801-250G 26-2801-1000G
101324-672EA 203.82 USD
101324-672 101324-670
Pentacarbonyliron ≥99.5% (99.9+%-Fe)
Pentacarbonyliron

Metal Carbonyls, Volatile Metal Carbonyls for CVD & ALD, Volatile Precursors for CVD

Formula: C₅FeO₅
Boiling Pt: 103 °C
Melting Pt: –20 °C
Density: 1.457 g/cm³ (20 °C)
Flash Pt: 5 °F
MDL Number: MFCD00011001
CAS Number: 13463-40-6
EINECS: 236-670-8
UN: 1994
ADR: 6.1,I
Merck Index: 12,05111

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR